missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2R1/CDC34 Antibody (CPTC-CDC34-2), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08839C
This item is not returnable.
View return policy
Description
UBE2R1/CDC34 Monoclonal antibody specifically detects UBE2R1/CDC34 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
| UBE2R1/CDC34 | |
| Monoclonal | |
| DyLight 650 | |
| cell division cycle 34, cell division cycle 34 homolog (S. cerevisiae), E2-CDC34, EC 6.3.2.19, UBC3, UBE2R1UBCH3, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 R1, Ubiquitin-conjugating enzyme E2-32 kDa complementing, Ubiquitin-conjugating enzyme E2-CDC34, Ubiquitin-protein ligase R1 | |
| Recombinant human full-length UBE2R1/CDC34 protein (Uniprot: P49427) | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 997 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| CPTC-CDC34-2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction