missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2S Antibody (OTI1B5), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74749AF532
This item is not returnable.
View return policy
Description
UBE2S Monoclonal specifically detects UBE2S in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| UBE2S | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| UBE2S | |
| Full length recombinant protein of human UBE2S (NP_055316) produced in E.coli. | |
| 0.1 mL | |
| 27338.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI1B5 | |
| Western Blot, Flow Cytometry | |
| E2-EPF5, E2-EPFEC 6.3.2.19, E2EPFEPF5, Ubiquitin carrier protein S, ubiquitin-conjugating enzyme E2 S, ubiquitin-conjugating enzyme E2-24 kD, Ubiquitin-conjugating enzyme E2-24 kDa, Ubiquitin-conjugating enzyme E2-EPF5, ubiquitin-conjugating enzyme E2S, Ubiquitin-protein ligase S | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction