Learn More
UBE2S Monoclonal Antibody (OTI4B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505169
Encompass_Preferred
 View more versions of this product
Product Code. 17563292
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
This gene encodes a member of the ubiquitin-conjugating enzyme family. The encoded protein is able to form a thiol ester linkage with ubiquitin in a ubiquitin activating enzyme-dependent manner, a characteristic property of ubiquitin carrier proteins.
Specifications
Specifications
UBE2S
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q16763
UBE2S
Full length human recombit protein of human UBE2S produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Western Blot
OTI4B8
Unconjugated
UBE2S
E2-EPF, E2EPF, EPF5
Mouse
Affinity Chromatography
RUO
27338
-20° C, Avoid Freeze/Thaw Cycles