missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2S Monoclonal Antibody (OTI4B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505169S
This item is not returnable.
View return policy
Description
UBE2S Monoclonal antibody specifically detects UBE2S in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| UBE2S | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16763 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 27338 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI4B8 | |
| Unconjugated | |
| UBE2S | |
| 0910001J09Rik; 6720465F12Rik; AA409170; E2 ubiquitin-conjugating enzyme S; E2EPF; E2-EPF; E2-EPF5; EPF5; OK/SW-cl.73; RGD1564746; UBE2S; Ubiquitin carrier protein S; ubiquitin conjugating enzyme E2 S; ubiquitin conjugating enzyme E2S; ubiquitin-conjugating enzyme E2 S; ubiquitin-conjugating enzyme E2-24 kD; ubiquitin-conjugating enzyme E2-24 kDa; Ubiquitin-conjugating enzyme E2-EPF5; ubiquitin-conjugating enzyme E2S; ubiquitin-protein ligase S | |
| Full length human recombinant protein of human UBE2S produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction