missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2S Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18219-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the ubiquitin-conjugating enzyme family. The encoded protein is able to form a thiol ester linkage with ubiquitin in a ubiquitin activating enzyme-dependent manner, a characteristic property of ubiquitin carrier proteins.Specifications
| UBE2S | |
| Polyclonal | |
| Unconjugated | |
| UBE2S | |
| E2 EPF, E2EPF, EPF5, UBE2S, Ubiquitin carrier protein S, Ubiquitin protein ligase S | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27338, 292588, 77891 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B5DFI8, Q16763, Q921J4 | |
| UBE2S | |
| UBE2S Fusion Protein Ag12978 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction