missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2T Antibody (OTI2F5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74750CL1
This item is not returnable.
View return policy
Description
UBE2T Monoclonal antibody specifically detects UBE2T in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| UBE2T | |
| Monoclonal | |
| CoraFluor 1 | |
| Cell proliferation-inducing gene 50 protein, EC 6.3.2.19, HSPC150, HSPC150 protein similar to ubiquitin-conjugating enzyme, PIG50, Ubiquitin carrier protein T, ubiquitin conjugating enzyme, ubiquitin-conjugating enzyme E2 T, ubiquitin-conjugating enzyme E2T (putative), Ubiquitin-protein ligase T | |
| Full length human recombinant protein of human UBE2T (NP_054895) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI2F5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 29089 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction