missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TR4/NR2C2 Antibody (SR2096), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22644-50ul
This item is not returnable.
View return policy
Description
TR4/NR2C2 Monoclonal antibody specifically detects TR4/NR2C2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TR4/NR2C2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR2096 | |
| Western Blot 1:500-1:2000 | |
| hTAK1, Nuclear hormone receptor TR4, nuclear receptor subfamily 2 group C member 2, nuclear receptor subfamily 2, group C, member 2, Orphan nuclear receptor TAK1, Orphan nuclear receptor TR4, TAK1TR4TR2R1, Testicular receptor 4, TR4 nuclear hormone receptor | |
| A synthesized peptide derived from human TR4/NR2C2 (Uniprot #: P49116) | |
| 50 μg | |
| Neuroscience | |
| 7182 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction