missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2T Antibody (SR1361), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | UBE2T |
|---|---|
| Clone | SR1361 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631105
|
Novus Biologicals
NBP3-22630-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681434
|
Novus Biologicals
NBP3-22630-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBE2T Monoclonal antibody specifically detects UBE2T in Human samples. It is validated for Western BlotSpecifications
| UBE2T | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 29089 | |
| IgG | |
| Affinity purified |
| SR1361 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Cell proliferation-inducing gene 50 protein, EC 6.3.2.19, HSPC150, HSPC150 protein similar to ubiquitin-conjugating enzyme, PIG50, Ubiquitin carrier protein T, ubiquitin conjugating enzyme, ubiquitin-conjugating enzyme E2 T, ubiquitin-conjugating enzyme E2T (putative), Ubiquitin-protein ligase T | |
| A synthesized peptide derived from human UBE2T (Uniprot #: Q9NPD8) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title