missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2U Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | UBE2U |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
UBE2U Polyclonal antibody specifically detects UBE2U in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| UBE2U | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 148581 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 6.3.2.19, MGC35130, RP4-636O23.1, Ubiquitin carrier protein U, ubiquitin-conjugating enzyme E2 U, ubiquitin-conjugating enzyme E2U (putative), Ubiquitin-protein ligase U | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-226 of human UBE2U (NP_689702.1). MHGRAYLLLHRDFCDLKENNYKGITAKPVSEDMMEWEVEIEGLQNSVWQGLVFQLTIHFTSEYNYAPPVVKFITIPFHPNVDPHTGQPCIDFLDNPEKWNTNYTLSSILLALQVMLSNPVLENPVNLEAARILVKDESLYRTILRLFNRPLQMKDDSQELPKDPRKCIRPIKTTSFSDYYQTWSRIATSKATEYYRTPLLKVPNFIGQYYKWKKMDLQHQKEWNLK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title