Learn More
Invitrogen™ UBE2V1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592896
Description
Immunogen sequence: MAATTGSGVK VPRNFRLLEE LEEGQKGVGD GTVSWGLEDD EDMTLTRWTG MIIGPPRTIY ENRIYSLKIE CGPKYPEAPP FVRFVTKINM NGVNSSNGVV DPRAISVLAK WQNSYSIKVV LQELRRLMMS KENMKLPQPP EGQCYSN; Positive Samples: BT-474, 293T, HepG2, HeLa, Mouse brain, Mouse pancreas, Mouse spinal cord, Rat liver, Rat spleen; Cellular Location: Nucleus.
The TMEM189-UEV mRNA is an infrequent but naturally occurring co-transcribed product of the neighboring TMEM189 and UBE2V1 genes. Ubiquitin-conjugating E2 enzyme variant proteins constitute a distinct subfamily within the E2 protein family. They have sequence similarity to other ubiquitin-conjugating enzymes but lack the conserved cysteine residue that is critical for the catalytic activity of E2s. The protein produced by this transcript has UEV1 B domains but the protein is localized to the cytoplasm rather than to the nucleus. The significance of this co-transcribed mRNA and the function of its protein product has not yet been determined.
Specifications
| UBE2V1 | |
| Polyclonal | |
| Unconjugated | |
| Ube2v1 | |
| 0610011J09Rik; AI256840; CIR1; CROC1; CROC-1; CROC1B; CROC-1B; CROC-1UBE2V; D7Bwg1382e; DJ1185N5.1.3; DNA-binding protein; KUA-UEV; P/OKcl.19; TRAF6-regulated IKK activator 1 beta Uev1A; UBE2V; Ube2v1; ubiquitin conjugating enzyme E2 V1; ubiquitin conjugating enzyme E2 variant 1; ubiquitin-conjugating enzyme E2 variant 1; UEV1; UEV-1; UEV1A; UEV-1CIR1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 296390, 66589, 7335 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13404, Q9CZY3 | |
| Ube2v1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human UBE2V1 (NP_0010274591). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.