missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2W Antibody (10), Alexa Fluor™ 350 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05884AF350
This item is not returnable.
View return policy
Description
UBE2W Monoclonal antibody specifically detects UBE2W in Human samples. It is validated for ELISA
Specifications
| UBE2W | |
| Monoclonal | |
| Alexa Fluor 350 | |
| EC 6.3.2.19, FLJ11011, hUBC-16, probable ubiquitin-conjugating enzyme E2 W, Ubiquitin carrier protein W, ubiquitin-conjugating enzyme 16, ubiquitin-conjugating enzyme E2W (putative), Ubiquitin-protein ligase W | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human UBE2W (Q96B02-12; Met1-Cys151). | |
| 0.1 mL | |
| Cell Biology | |
| 55284 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction