Learn More
Invitrogen™ UBE2Z Monoclonal Antibody (K52034_14C7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556285
Description
Sequence of this protein is as follows: WDGERTAPQC LLRIKRDIMS IYKEPPPGMF VVPDTVDMTK IHALITGPFD TPYEGGFFLF VFRCPPDYPI HPPRVKLMTT GNNTVRFNPN FYRNGKVCLS ILGTWTGPAW SPAQSISSVL ISIQSLMTEN PYHNEPGFEQ ERHPGDSKNY NECIRHETIR VAVCDMMEGK CPCPEPLRGV MEKSFLEYYD FYEVACKDRL HLQGQTMQDP FGEKRGHFDY QSLLMRLGLI RQKVLERLHN ENAEMDSDSS
This gene encodes an enzyme which ubiquitinates proteins which participate in signaling pathways and apoptosis.
Specifications
| UBE2Z | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9H832 | |
| UBE2Z | |
| Recombinant human UBE2Z (immunogen range is 91-340). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K52034_14C7 | |
| Unconjugated | |
| UBE2Z | |
| AW049199; C030047H17Rik; D11Moh35; E2 ubiquitin-conjugating enzyme Z; HOYS7; RGD1308347; uba6-specific E2 conjugating enzyme 1; UBA6-specific enzyme E2; UBE2Z; Ubiquitin carrier protein Z; ubiquitin conjugating enzyme E2 Z; ubiquitin conjugating enzyme E2Z; ubiquitin-conjugating enzyme E2 Z; ubiquitin-conjugating enzyme E2Z; ubiquitin-conjugating enzyme E2Z (putative); ubiquitin-protein ligase Z; Use1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 65264 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.