missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBE2Z Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551237
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total UBE2Z protein.
This gene encodes an enzyme which ubiquitinates proteins which participate in signaling pathways and apoptosis.
Specifications
| UBE2Z | |
| Polyclonal | |
| Unconjugated | |
| UBE2Z | |
| AW049199; C030047H17Rik; D11Moh35; E2 ubiquitin-conjugating enzyme Z; HOYS7; RGD1308347; uba6-specific E2 conjugating enzyme 1; UBA6-specific enzyme E2; UBE2Z; Ubiquitin carrier protein Z; ubiquitin conjugating enzyme E2 Z; ubiquitin conjugating enzyme E2Z; ubiquitin-conjugating enzyme E2 Z; ubiquitin-conjugating enzyme E2Z; ubiquitin-conjugating enzyme E2Z (putative); ubiquitin-protein ligase Z; Use1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 268470, 303478, 65264 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q3B7D1, Q3UE37, Q9H832 | |
| UBE2Z | |
| Fusion protein of human UBE2Z. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction