missing translation for 'onlineSavingsMsg'
Learn More

UBE3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16866673 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16866673 20 μL 20µL
16856673 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16866673 Supplier Proteintech Supplier No. 103441AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The E6-associated protein (E6-AP/UBE3A), a 100a protein, was originally identified as an ubiquitin-protein ligase (E3), in the presence of the E6 protein from human papillomavirus types 16 and 18. E6-AP has also been shown to function as a coactivator of steroid receptor-dependent transcriptional activity. Recent data indicate that the ubiquitin-protein ligase function of E6-AP is dispensable for its ability to coactivate nuclear hormone receptors, showing that E6-AP possesses two separable independent functions, as both a coactivator and a ubiquitin-protein ligase.

Specifications

Antigen UBE3A
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBE3A
Gene Accession No. O08759, Q05086
Gene Alias ANCR, AS, E6 AP, E6AP, E6AP ubiquitin protein ligase, EPVE6AP, HPVE6A, UBE3A, ubiquitin protein ligase E3A
Gene Symbols Ube3a
Host Species Rabbit
Immunogen UBE3A Fusion Protein Ag0346
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22215, 361585, 7337
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.