missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE3B Monoclonal Antibody (OTI3F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806725
This item is not returnable.
View return policy
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: E1 ubiquitin-activating enzymes, E2 ubiquitin-conjugating enzymes, and E3 ubiquitin-protein ligases. This gene encodes a member of the E3 ubiquitin-conjugating enzyme family which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme and transfers the ubiquitin to the targeted substrates. A HECT (homology to E6-AP C-terminus) domain in the C-terminus of the longer isoform of this protein is the catalytic site of ubiquitin transfer and forms a complex with E2 conjugases. Shorter isoforms of this protein which lack the C-terminal HECT domain are therefore unlikely to bind E2 enzymes. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.Specifications
| UBE3B | |
| Monoclonal | |
| Unconjugated | |
| UBE3B | |
| BPIDS, KOS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 89910 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| OTI3F10 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z3V4 | |
| UBE3B | |
| Human recombit protein fragment corresponding to amino acids 768-1068 of human UBE3B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction