missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE4B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98954-100ul
This item is not returnable.
View return policy
Description
UBE4B Polyclonal antibody specifically detects UBE4B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| UBE4B | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| E4, homologous to yeast UFD2, Homozygously deleted in neuroblastoma 1, homozygously deleted in neuroblastoma-1, KIAA0684HDNB1, ubiquitin conjugation factor E4 B, Ubiquitin fusion degradation protein 2, ubiquitination factor E4B (homologous to yeast UFD2), ubiquitination factor E4B (UFD2 homolog, yeast), ubiquitin-fusion degradation protein 2, UBOX3, UFD2UFD2A | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human UBE4B. | |
| 100 μg | |
| Apoptosis | |
| 10277 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction