missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBIAD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 470.00
Specifications
| Antigen | UBIAD1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697840
|
Novus Biologicals
NBP2-93402-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18647791
|
Novus Biologicals
NBP2-93402-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UBIAD1 Polyclonal antibody specifically detects UBIAD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| UBIAD1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 29914 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.5.1.-, RP4-796F18.1, SCCD, Schnyder crystalline corneal dystrophy, TERE1transitional epithelia response protein, Transitional epithelial response protein 1, UbiA prenyltransferase domain containing 1, ubiA prenyltransferase domain-containing protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human UBIAD1 (NP_037451.1). PEQDRLPQRSWRQK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title