missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBIAD1 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-468IR
This item is not returnable.
View return policy
Description
UBIAD1 Polyclonal antibody specifically detects UBIAD1 in Human samples. It is validated for Western Blot
Specifications
| UBIAD1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 755 | |
| EC 2.5.1.-, RP4-796F18.1, SCCD, Schnyder crystalline corneal dystrophy, TERE1transitional epithelia response protein, Transitional epithelial response protein 1, UbiA prenyltransferase domain containing 1, ubiA prenyltransferase domain-containing protein 1 | |
| A synthetic peptide made to residues 32-45 [PEQDRLPQRSWRQK] of the 37 kDa human TERE1 protein. | |
| 0.1 mL | |
| Cancer | |
| 29914 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction