missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ubinuclein 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571822
This item is not returnable.
View return policy
Description
Ubinuclein 2 Polyclonal Antibody for Flow, Western Blot
Ubinuclein 1 (UBN1) is a ubiquitously expressed evolutionarily conserved protein which binds to proliferation-promoting genes that are repressed by formation of senescence-associated heterochromatin foci (SAHF). Ubinuclein 1 associates with various transcription factors and with histone methyltransferase activity, is indispensable for SAHF formation and appears to be a regulator of senescence. Although in most cells ubinuclein is localized to the nucleus, in cells forming tight junctions it is recruited to the cell adhesion complexes, dependently on the cell density.
Specifications
| Ubinuclein 2 | |
| Polyclonal | |
| Unconjugated | |
| UBN2 | |
| 2900060J04Rik; 6030408G03Rik; BC060116; D130059P03Rik; Kiaa2030; mKIAA2030; ubinuclein 1; ubinuclein 2; ubinuclein-2; UBN1; UBN2; VT; VT4 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 254048, 320538 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6ZU65, Q80WC1 | |
| UBN2 | |
| KLH conjugated synthetic peptide between 171-200 amino acids from the N-terminal region of human UBN2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction