missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ubiquilin 2 Monoclonal Antibody (6H9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547466
This item is not returnable.
View return policy
Description
Ubiquilin 2 Monoclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases; and thus, are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to bind the ATPase domain of the Hsp70-like Stch protein.
Specifications
| Ubiquilin 2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5 mM sodium azide | |
| Q9QZM0, Q9UHD9 | |
| UBQLN2 | |
| Human ubiquilin 2 expressed in and purified from E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 6H9 | |
| Unconjugated | |
| UBQLN2 | |
| ALS15; CHAP1; DSK2; DSK2 homolog; HRIHFB2157; N4BP4; Nedd4 binding protein 4; Plic2; Plic-2; protein linking IAP with cytoskeleton 2; RGD1563566; ubiquilin 2; ubiquilin-2; Ubiquitin-like product Chap1/Dsk2; Ubiquitin-related protein; Ubqln2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 29978, 317396, 54609 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction