missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ubiquilin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106760
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total UBQLN3.
This gene encodes an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This gene is specifically expressed in the testis, and proposed to regulate cell-cycle progression during spermatogenesis.
Specifications
| Ubiquilin 3 | |
| Polyclonal | |
| Unconjugated | |
| UBQLN3 | |
| 4933400K24Rik; testicular tissue protein Li 220; TUP-1; ubiquilin 3; ubiquilin-3; Ubqln3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 244178, 308905, 50613 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8C5U9, Q9H347 | |
| UBQLN3 | |
| A synthesized peptide derived from human UBQLN3(Accession Q9H347), corresponding to amino acid residues F273-D323. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction