missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin Antibody (Ubi-1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-130AF594
This item is not returnable.
View return policy
Description
Ubiquitin Monoclonal antibody specifically detects Ubiquitin in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Drosophila, Equine, Plant, Monkey, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Specifications
| Ubiquitin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, C. elegans, Chicken, Drosophila, Equine, Plant, Monkey, Zebrafish | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
| Ubi-1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting | |
| RPS27A, UBA52, UBB ubiquitin B, UBC | |
| Purified bovine erythrocyte ubiquitin cross-linked with glutaraldehyde to KLH | |
| 0.1 mL | |
| Alzheimers Research, Autophagy, Cancer, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Turnover, Ubiquitin Proteasome Pathway | |
| 6233 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction