missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ubiquitin B Antibody (6C11-B3), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12882V
This item is not returnable.
View return policy
Description
Ubiquitin B Monoclonal specifically detects Ubiquitin B in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Ubiquitin B | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| UBB | |
| Native bovine ubiquitin, conjugated to KLH | |
| 200 μL | |
| 7314 | |
| Human, Mouse, Rat, Bovine | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 6C11-B3 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects approx 10kDa. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction