missing translation for 'onlineSavingsMsg'
Learn More

Ubiquitin B Antibody (UBB/2122), HRP, Novus Biologicals™

Product Code. 18650047 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18650047 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18650047 Supplier Novus Biologicals Supplier No. NBP308906H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ubiquitin B Monoclonal antibody specifically detects Ubiquitin B in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin B
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone UBB/2122
Conjugate HRP
Formulation PBS
Gene Alias FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B
Host Species Mouse
Immunogen Recombinant fragment of human Ubiquitin B protein (around aa 1-119) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7314
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.