missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBL3 Antibody (OTI1G4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74751B
This item is not returnable.
View return policy
Description
UBL3 Monoclonal specifically detects UBL3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| UBL3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| UBL3 | |
| Recombinant protein fragment corresponding to amino acids 1-114 of human UBL3 (NP_009037) produced in E.coli. | |
| 0.1 mL | |
| 5412.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1G4 | |
| Western Blot | |
| DKFZP434K151, FLJ32018, HCG-1, hsMUB, Membrane-anchored ubiquitin-fold protein, MUB, PNSC1, Protein HCG-1, ubiquitin-like 3, ubiquitin-like protein 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction