missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBL4A Antibody (OTI1D4), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74752AF647
This item is not returnable.
View return policy
Description
UBL4A Monoclonal specifically detects UBL4A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| UBL4A | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| UBL4A | |
| Full length recombinant protein of human UBL4A (NP_055050) produced in E.coli. | |
| 0.1 mL | |
| 8266.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1D4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DXS254EUbiquitin-like protein GDX, G6PD, GDXubiquitin-like 4, GET5, MDY2, TMA24, ubiquitin-like 4A, ubiquitin-like protein 4A, UBL4DX254E | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction