missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBL4B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551149
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total UBL4B protein.
Ubiquitin is a 77 amino acid protein that targets proteins for degradation by the 26S proteasome. Ubiquitin-like proteins are not directly involved in protein degradation, but appear to have many mechanistic similarities with the ubiquitin pathway. UBL4B (Ubiquitin-like protein 4B) is a 174 amino acid cytoplasmic protein that contains a ubiquitin-like domain. UBL4B likely arose by retroposition during mammalian evolution from UBL4A, an X-linked intron-bearing housekeeping gene. While UBL4A is highly conserved, UBL4B has undergone rapid evolution and may have evolved new functions. Expression of UBL4B is restricted to post-meiotic germ cells in testis and ovarian tissue, where it likely functions in post-translational protein modification.
Specifications
| UBL4B | |
| Polyclonal | |
| Unconjugated | |
| UBL4B | |
| 4930522D07Rik; FLJ25690; NEWGENE_1589125; ubiquitin like 4B; ubiquitin-like 4B; ubiquitin-like protein 4B; UBL4B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 164153 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8N7F7 | |
| UBL4B | |
| Full length fusion protein of human UBL4B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction