missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBN1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-26142
This item is not returnable.
View return policy
Description
UBN1 Polyclonal specifically detects UBN1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| UBN1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| UBN1 | |
| Peptide with sequence C-DNSEAYDELVPASLT corresponding to internal region according to NP_058632.2. | |
| 0.1 mg | |
| Primary | |
| Reported variants represent identical protein: NP_058632.2, NP_001072982.1. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:128000 | |
| HIRA-binding protein, ubinuclein 1, ubinuclein-1, VT, VT4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29855 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction