missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBN1 Antibody (UBN1-02), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62205V
This item is not returnable.
View return policy
Description
UBN1 Monoclonal specifically detects UBN1 in Human samples. It is validated for Western Blot.
Specifications
| UBN1 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| UBN1 | |
| Recombinant protein corresponding to amino acids 1-190 of ubinuclein 1. | |
| 0.1 mL | |
| 29855 | |
| Human | |
| IgG |
| Western Blot | |
| UBN1-02 | |
| Western Blot | |
| HIRA-binding protein, ubinuclein 1, ubinuclein-1, VT, VT4 | |
| Mouse | |
| Protein A purified | |
| Primary | |
| The antibody SC-05 reacts with 80 kDa human secretory component glycoprotein (both free and bound in secretory IgA). Recognized glycoprotein is specific membrane marker of glandular carcinomas. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction