missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBOX5 Antibody (OTI8D5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74757H
This item is not returnable.
View return policy
Description
UBOX5 Monoclonal specifically detects UBOX5 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| UBOX5 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| UBOX5 | |
| Recombinant protein fragment corresponding to amino acids 1-130 and 419-487 of human UBOX5 (NP_955447) produced in E.coli. | |
| 0.1 mL | |
| 22888.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI8D5 | |
| Western Blot, Immunohistochemistry | |
| KIAA0860U-box domain-containing protein 5, RING finger protein 37, RNF37Ubce7ip5, UBCE7IP5, ubiquitin conjugating enzyme 7 interacting protein 5, Ubiquitin-conjugating enzyme 7-interacting protein 5, U-box domain containing 5, UIP5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction