missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBOX5 Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503340
This item is not returnable.
View return policy
Description
UBOX5 encodes a U-box domain containing protein. The encoded protein interacts with E2 enzymes and may play a role in the ubiquitination pathway. Alternate splicing results in multiple transcript variants.Specifications
| UBOX5 | |
| Monoclonal | |
| Unconjugated | |
| UBOX5 | |
| RNF37, UBCE7IP5, UIP5, hUIP5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 22888 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O94941 | |
| UBOX5 | |
| Human recombit protein fragment corresponding to amino acids 1-130 and C 419-487 of human UBOX5 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction