missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBOX5 Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503340S
This item is not returnable.
View return policy
Description
UBOX5 Monoclonal antibody specifically detects UBOX5 in Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| UBOX5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| B5DEQ3, B7ZMP1, Q9NQH7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 321003, 481237, 63929, 685823 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C5 | |
| Unconjugated | |
| XPNPEP3 | |
| 1500010O06Rik; C330018L13Rik; FAST kinase domain-containing protein 5; RING finger protein 37; FAST kinase domains 5; FASTKD5; hUIP5; KIAA0860; RGD1305440; RING finger protein 37; RING-type E3 ubiquitin transferase RNF37; Rnf37; U box domain containing 5; UBCE7IP5; ubcM4-interacting protein 5; Ube7ip5; ubiquitin conjugating enzyme 7 interacting protein 5; ubiquitin-conjugating enzyme 7-interacting protein 5; U-box domain containing 5; U-box domain-containing protein 5; Ubox5; UIP5; Unknown (protein for MGC:138103) | |
| Full length human recombinant protein of human XPNPEP3 produced in HEK293 cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction