Learn More
UBOX5 Mouse anti-Human, Clone: OTI1C5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503340
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
UBOX5 encodes a U-box domain containing protein. The encoded protein interacts with E2 enzymes and may play a role in the ubiquitination pathway. Alternate splicing results in multiple transcript variants.Specifications
| UBOX5 | |
| Monoclonal | |
| Unconjugated | |
| UBOX5 | |
| RNF37, UBCE7IP5, UIP5, hUIP5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 22888 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O94941 | |
| UBOX5 | |
| Human recombit protein fragment corresponding to amino acids 1-130 and C 419-487 of human UBOX5 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.