missing translation for 'onlineSavingsMsg'
Learn More

UBP1 Monoclonal Antibody (OTI9B3), TrueMAB™, OriGene

Product Code. 17500554
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17500554 30 μL 30µL
17580544 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17500554 Supplier Origene Technologies Supplier No. TA808852S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBP1 Monoclonal antibody specifically detects UBP1 in Human samples. It is validated for Western Blot

Specifications

Antigen UBP1
Applications Western Blot
Classification Monoclonal
Clone OTI9B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene UBP1
Gene Accession No. Q9NZI7
Gene Alias Cp2b; LBP1; LBP1A; LBP-1a; LBP1B; LBP-1b; NF2d9; Nuclear factor 2d9; RGD1564700; transcription factor LBP-1; Ubp1; upstream binding protein 1; upstream binding protein 1 (LBP-1a); upstream-binding protein 1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 47-269 of human UBP1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7342
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.