missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBP1 Monoclonal Antibody (OTI9B3), TrueMAB™, OriGene
Description
UBP1 functions as a transcriptional activator in a promoter context-dependent manner. Modulates the placental expression of CYP11A1. Involved in regulation of the alpha-globin gene in erythroid cells. Activation of the alpha-globin promoter in erythroid cells is via synergistic interaction with TFCP2. Involved in regulation of the alpha-globin gene in erythroid cells. Binds strongly to sequences around the HIV-1 initiation site and weakly over the TATA-box. Represses HIV-1 transcription by inhibiting the binding of TFIID to the TATA-box.
Specifications
Specifications
| Antigen | UBP1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI9B3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | UBP1 |
| Gene Accession No. | Q9NZI7 |
| Gene Alias | LBP-1B, LBP-1a, LBP1A, LBP1B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?