missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBPY/USP8 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94523-0.1ml
This item is not returnable.
View return policy
Description
UBPY/USP8 Polyclonal antibody specifically detects UBPY/USP8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| UBPY/USP8 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Deubiquitinating enzyme 8, EC 3.1.2.15, EC 3.4.19.12, HumORF8, KIAA0055hUBPy, MGC129718, ubiquitin carboxyl-terminal hydrolase 8, Ubiquitin isopeptidase Y, ubiquitin specific peptidase 8, ubiquitin specific protease 8, ubiquitin thioesterase 8, Ubiquitin thiolesterase 8, Ubiquitin-specific-processing protease 8, UBPYFLJ34456 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human USP8 (NP_005145.3). MPAVASVPKELYLSSSLKDLNKKTEVKPEKISTKSYVHSALKIFKTAEECRLDRDEERAYVLYMKYVTVYNLIKKRPDFKQQQDYFHSILGPGNIKKAVEEAERLSESLKLRYEEAEVRKKLEEKDRQEEAQRLQQKRQETGREDGGTLAKGSLENVLDSKDKTQKSNGEKNEKCETKEKGAITAKELYTMMTDKNISLIIMDARRMQDYQDSCILHSLSVPEEAISPGVTASWIEAHLPDDSKDTWKKRGNVEYVVLLDWFSSAKDLQI | |
| 0.1 mL | |
| Cell Biology | |
| 9101 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction