missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBQLN3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93521-0.1ml
This item is not returnable.
View return policy
Description
UBQLN3 Polyclonal antibody specifically detects UBQLN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| UBQLN3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| TUP-1, ubiquilin 3, ubiquilin-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 306-620 of human UBQLN3 (NP_059509.1). WTSTHGGSGSRQGRQDGDQDAPDIRNRFPNFLGIIRLYDYLQQLHENPQSLGTYLQGTASALSQSQEPPPSVNRVPPSSPSSQEPGSGQPLPEESVAIKGRSSCPAFLRYPTENSTGQGGDQDGAGKSSTGHSTNLPDLVSGLGDSANRVPFAPLSFSPTAAIPGIPEPPWLPSPAYPRSLRPDGMNPAPQLQDEIQPQLPLLMHLQAAMANPRALQALRQIEQGLQVLATEAPRLLLWFMPCLAGTGSVAGGIESREDPLMSEDPLPNPPPEVFPALDSAELGFLSPPFLHMLQDLVSTNPQQLQPEAHFQVQL | |
| 0.1 mL | |
| Cell Biology | |
| 50613 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction