missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBR5/EDD Antibody (SR1501), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22217-50ul
This item is not returnable.
View return policy
Description
UBR5/EDD Monoclonal antibody specifically detects UBR5/EDD in Human, Mouse samples. It is validated for Western Blot
Specifications
| UBR5/EDD | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| SR1501 | |
| Western Blot 1:500-1:2000 | |
| DD5, E3 ubiquitin protein ligase, HECT domain containing, 1, E3 ubiquitin-protein ligase, HECT domain-containing 1, EC 6.3.2, EC 6.3.2.-, EDD1E3 ubiquitin-protein ligase UBR5, EDDE3 identified by differential display, hHYD, HYDFLJ11310, Hyperplastic discs protein homolog, KIAA0896MGC57263, progestin induced protein, Progestin-induced protein, ubiquitin protein ligase E3 component n-recognin 5, ubiquitin-protein ligase | |
| A synthesized peptide derived from human UBR5/EDD (Uniprot #: O95071) | |
| 50 μg | |
| DNA Repair | |
| 51366 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto