missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UBTD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557449
This item is not returnable.
View return policy
Description
Immunogen sequence: QLPVYCLAPP INMIEEKSDI ETLDIPEPPP NSGYECQLRL RLSTGKDLKL VVRSTD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
UBTD2 gene ontology (GO) annotation includes cytoplasm and cytoplasm.
Specifications
| UBTD2 | |
| Polyclonal | |
| Unconjugated | |
| UBTD2 | |
| 4930571L24; 9630054F20Rik; AI645720; DCUBP; DC-UbP; Dendritic cell-derived ubiquitin-like protein; RGD1305506; SB72; ubiquitin domain containing 2; ubiquitin domain-containing protein 2; Ubiquitin-like protein SB72; Ubtd2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 92181 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WUN7 | |
| UBTD2 | |
| Recombinant protein corresponding to Human UBTD2. Recombinant protein control fragment (Product #RP-102686). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction