Learn More
UBXD1 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL594-14706-100UL
Description
The UBXD1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human UBXD1. This antibody recognizes human, mouse, rat antigen. The UBXD1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| UBXD1 | |
| Polyclonal | |
| CoraLite 594 | |
| 100 μL | |
| 80700 | |
| Liquid |
| Immunofluorescence | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| IgG |