missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXD2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18404IR
This item is not returnable.
View return policy
Description
UBXD2 Polyclonal antibody specifically detects UBXD2 in Human samples. It is validated for Western Blot
Specifications
| UBXD2 | |
| Polyclonal | |
| Western Blot | |
| erasin, FLJ23318, KIAA0242KIAA2042, UBX domain containing 2, UBX domain protein 4, UBX domain-containing 1, UBX domain-containing 2, UBX domain-containing protein 2, UBX domain-containing protein 4, UBXD2, UBXDC1 | |
| Recognizes region between residues Between Between 458 and 508.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23190 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction