Learn More
UBXN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12567-50ul
This item is not returnable.
View return policy
Description
UBXN1 Polyclonal antibody specifically detects UBXN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)