missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXN2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98564-50ul
This item is not returnable.
View return policy
Description
UBXN2A Polyclonal antibody specifically detects UBXN2A in Human samples. It is validated for Western Blot
Specifications
| UBXN2A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC138202, UBX domain containing 4, UBX domain protein 2A, UBX domain-containing protein 2A, UBX domain-containing protein 4, UBXD4 | |
| Produced in rabbits immunized with E. coli-derived Human UBXN2A fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 165324 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction