missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXN2B Antibody (OTI1F7), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74769IR
This item is not returnable.
View return policy
Description
UBXN2B Monoclonal specifically detects UBXN2B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| UBXN2B | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| UBXN2B | |
| Full length recombinant protein of human UBXN2B (NP_001071087) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1F7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| UBX domain protein 2B | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 137886.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction