missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXN2B Antibody (OTI1F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00543
This item is not returnable.
View return policy
Description
UBXN2B Monoclonal specifically detects UBXN2B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| UBXN2B | |
| Monoclonal | |
| 0.41 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
| UBXN2B | |
| Full length human recombinant protein of human UBXN2B (NP_001071087) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI1F7 | |
| Unconjugated | |
| UBX domain protein 2B | |
| Mouse | |
| 36.9 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 137886 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction