missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBXN2B Mouse anti-Canine, Human, Clone: OTI3H8, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502539
This item is not returnable.
View return policy
Description
UBXD2B (UBX domain-containing protein 2B), also known as NSFL1 cofactor p37 and p97 cofactor p37, is a 331 amino acid protein that contains one UBX domain and one SEP domain. UBXN2B is required for ER and Golgi biogenesis and also plays a role in their maintece during interphase, as well as their reassembly at the end of mitosis. Through interaction with VCP, UBXN2B forms a complex that has membrane fusion activity. Adapter protein required for Golgi and endoplasmic reticulum biogenesis. Involved in Golgi and endoplasmic reticulum maintece during interphase and in their reassembly at the end of mitosis. The complex formed with VCP has membrane fusion activity; membrane fusion activity requires USO1-GOLGA2 tethering and BET1L. VCPIP1 is also required, but not its deubiquitinating activity.Specifications
| UBXN2B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14CS0 | |
| UBXN2B | |
| Full length human recombit protein of human UBXN2B produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI3H8 | |
| Unconjugated | |
| UBXN2B | |
| p37 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 137886, 477891 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction