missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ UBXN8 Antibody [DyLight 594], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-26909DL594
This item is not returnable.
View return policy
Description
UBXN8 Polyclonal antibody specifically detects UBXN8 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| UBXN8 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 7993 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| D8S2298EUBXD6, Rep-8 protein, REP8UBX domain-containing protein 6, Reproduction 8 protein, Reproduction/chromosome 8, UBX domain containing 6, UBX domain protein 8, UBX domain-containing protein 8 | |
| Produced in rabbits immunized with E. coli-derived Human UBXN8 fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction