missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCH-L1 Mouse anti-Human, Clone: 972106, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB60071-100UG
This item is not returnable.
View return policy
Description
UCH-L1/PGP9.5 Monoclonal specifically detects UCH-L1/PGP9.5 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| UCH-L1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| UCHL1 | |
| E. coli-derived recombinant human UCH-L1/PGP9.5 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 972106 | |
| Western Blot, Simple Western, Immunohistochemistry | |
| EC 3.4.19.12, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCH-L1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 7345 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction