Learn More
UCH-L1/PGP9.5 Antibody (13C4), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP2-29420-20ug
 View more versions of this product
Product Code. 18494402
  • € 291.00 / 20µg
Description
UCH-L1/PGP9.5 Monoclonal specifically detects UCH-L1/PGP9.5 in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Rabbit, Sheep, Zebrafish samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UCH-L1/PGP9.5
Monoclonal
0.2 mg/ml
Western Blot 0.5-1 ug/mL, Simple Western 10 ug/mL, ELISA 1-5 ug/mL for coating, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Frozen 0.5-1.0 ug/mL
EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1
Mouse
Protein A or G purified
RUO
Primary
This MAb reacts with a protein of 20-30kDa, identified as PGP9.5, also known as ubiquitin carboxyl-terminal hydrolase-1 (UchL1). Initially, PGP9.5 expression in normal tissues was reported in neurons and neuroendocrine cells but later it was found in distal renal tubular epithelium, spermatogonia, Leydig cells, oocytes, melanocytes, prostatic secretory epithelium, ejaculatory duct cells, epididymis, mammary epithelial cells, Merkel cells, and dermal fibroblasts. Furthermore, immunostaining for PGP9.5 has been shown in a wide variety of mesenchymal neoplasms as well. A mutation in PGP9.5 gene is believed to cause a form of Parkinson's disease.
Store at 4C.
IgG2a κ
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
13C4
Unconjugated
P09936
UCHL1
UCH-L1/PGP9.5 protein from brain (Uniprot: P09936)
20 ug
Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission
7345
Human, Mouse, Rat, Pig, Bovine, Canine, Guinea Pig, Rabbit, Sheep, Zebrafish