Learn More
UCH-L1/PGP9.5 Antibody (BH7), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-58869PCP
This item is not returnable.
View return policy
Description
UCH-L1/PGP9.5 Monoclonal antibody specifically detects UCH-L1/PGP9.5 in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry Free-Floating
Specifications
| UCH-L1/PGP9.5 | |
| Recombinant full length human UCHL1 purified from E. coli. [Swiss-Prot# P09936] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Pig, Bovine |
| Western Blot, Immunohistochemistry | |
| Affinity purified | |
| RUO | |
| 7345 | |
| IgG1 |